Assay Details
Gene Symbol: SS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198036.2 | NP_001184965.1 | 14 - 15 | 3260 | 75 | |
NM_101044.3 | NP_172637.2 | 13 - 14 | 3149 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
