Assay Details
Gene Symbol: SS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198036.2 | NP_001184965.1 | 3 - 4 | 1553 | 76 | |
NM_101044.3 | NP_172637.2 | 2 - 3 | 1442 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
