Assay Details
| Gene Symbol: KEU | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101108.4 | NP_563905.1 | 10 - 11 | 1191 | 124 | |
| GenBank mRNA | AK227584.1 | - | 10 - 11 | 1179 | 124 | |
More Information
Set Membership: |
|

KEU
Genomic Map