Assay Details
| Gene Symbol: KEU | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101108.4 | NP_563905.1 | 19 - 20 | 2166 | 108 | |
| GenBank mRNA | AK227584.1 | - | 19 - 20 | 2154 | 108 | |
More Information
Set Membership: |
|

KEU
Genomic Map