Assay Details
| Gene Symbol: KEU | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101108.4 | NP_563905.1 | 8 - 9 | 910 | 64 | |
| GenBank mRNA | AK227584.1 | - | 8 - 9 | 898 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

KEU| Gene Symbol: KEU | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101108.4 | NP_563905.1 | 8 - 9 | 910 | 64 | |
| GenBank mRNA | AK227584.1 | - | 8 - 9 | 898 | 64 | |
Set Membership: |
|