Assay Details
Gene Symbol: CLP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101113.4 | NP_563907.1 | 3 - 4 | 239 | 89 | |
GenBank mRNA | AB022327.1 | - | 3 - 4 | 208 | 89 | |
AY062770.1 | - | 3 - 4 | 217 | 89 | ||
AY081641.1 | - | 3 - 4 | 170 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
