Assay Details
Gene Symbol: CLP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101113.4 | NP_563907.1 | 6 - 7 | 604 | 88 | |
GenBank mRNA | AB022327.1 | - | 6 - 7 | 573 | 88 | |
AY062770.1 | - | 6 - 7 | 582 | 88 | ||
AY081641.1 | - | 6 - 7 | 535 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
