Assay Details
Gene Symbol: CLP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101113.4 | NP_563907.1 | 7 - 8 | 671 | 125 | |
GenBank mRNA | AB022327.1 | - | 7 - 8 | 640 | 125 | |
AY062770.1 | - | 7 - 8 | 649 | 125 | ||
AY081641.1 | - | 7 - 8 | 602 | 125 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
