Assay Details
Gene Symbol: CLP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101113.4 | NP_563907.1 | 8 - 9 | 769 | 131 | |
GenBank mRNA | AB022327.1 | - | 8 - 9 | 738 | 131 | |
AY062770.1 | - | 8 - 9 | 747 | 131 | ||
AY081641.1 | - | 8 - 9 | 700 | 131 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
