Assay Details
| Gene Symbol: ACR8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332035.1 | NP_001321345.1 | 1 - 2 | 141 | 150 | |
| NM_101114.4 | NP_172704.1 | 2 - 3 | 398 | 150 | ||
| GenBank mRNA | AF528064.1 | - | 2 - 3 | 142 | 150 | |
| AK228687.1 | - | 2 - 3 | 315 | 150 | ||
| BT026518.1 | - | 2 - 3 | 104 | 150 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001332035.1 transcripts.
Set Membership: |
|

ACR8
Genomic Map