Assay Details
| Gene Symbol: ACR8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332035.1 | NP_001321345.1 | 4 - 5 | 902 | 79 | |
| NM_101114.4 | NP_172704.1 | 5 - 6 | 1159 | 79 | ||
| GenBank mRNA | AF528064.1 | - | 5 - 6 | 903 | 79 | |
| AK228687.1 | - | 5 - 6 | 1076 | 79 | ||
| BT026518.1 | - | 5 - 6 | 865 | 79 | ||
More Information
Set Membership: |
|

ACR8
Genomic Map