Assay Details
Gene Symbol: ARK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198043.1 | NP_001184972.1 | 12 - 13 | 2195 | 78 | |
NM_101115.3 | NP_563908.1 | 12 - 13 | 2226 | 78 | ||
GenBank mRNA | AB290930.1 | - | 12 - 13 | 2000 | 78 | |
AF159052.1 | - | 12 - 13 | 2190 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
