Assay Details
Gene Symbol: ARK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198043.1 | NP_001184972.1 | 1 - 2 | 412 | 81 | |
NM_101115.3 | NP_563908.1 | 1 - 2 | 443 | 81 | ||
GenBank mRNA | AB290930.1 | - | 1 - 2 | 217 | 81 | |
AF159052.1 | - | 1 - 2 | 407 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
