Assay Details
Gene Symbol: ACA.l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101192.3 | NP_172780.1 | 10 - 11 | 3199 | 75 | |
GenBank mRNA | AY099862.1 | - | 10 - 11 | 3127 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
