Assay Details
Gene Symbol: HRS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101201.2 | NP_563926.1 | 1 - 2 | 248 | 75 | |
GenBank mRNA | AF367336.1 | - | 1 - 2 | 236 | 75 | |
AY124865.1 | - | 1 - 2 | 149 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
