Assay Details
| Gene Symbol: ELF5A-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101261.3 | NP_172848.1 | 4 - 5 | 572 | 89 | |
| GenBank mRNA | AY063780.1 | - | 4 - 5 | 415 | 89 | |
| AY117272.1 | - | 4 - 5 | 375 | 89 | ||
More Information
Set Membership: |
|

ELF5A-1
Genomic Map