Assay Details
| Gene Symbol: AT1G14160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101281.3 | NP_172868.1 | 1 - 2 | 476 | 86 | |
| GenBank mRNA | AK117303.1 | - | 1 - 2 | 476 | 86 | |
| BT006211.1 | - | 1 - 2 | 288 | 86 | ||
More Information
Set Membership: |
|

AT1G14160
Genomic Map