Assay Details
| Gene Symbol: FLP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084065.3 | NP_001077534.1 | 3 - 4 | 558 | 125 | |
| NM_101302.3 | NP_563948.1 | 2 - 3 | 323 | 125 | ||
| GenBank mRNA | AF371982.2 | - | 2 - 3 | 373 | 125 | |
| BT026126.1 | - | 2 - 3 | 145 | 125 | ||
More Information
Set Membership: |
|

FLP
Genomic Map