Assay Details
| Gene Symbol: FLP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084065.3 | NP_001077534.1 | 8 - 9 | 1198 | 152 | |
| NM_001332112.1 | NP_001321792.1 | 7 - 8 | 1166 | 152 | ||
| NM_001332113.1 | NP_001321793.1 | 6 - 7 | 955 | 152 | ||
| NM_101302.3 | NP_563948.1 | 7 - 8 | 963 | 152 | ||
| GenBank mRNA | AF371982.2 | - | 7 - 8 | 1013 | 152 | |
| BT026126.1 | - | 7 - 8 | 785 | 152 | ||
More Information
Set Membership: |
|

FLP
Genomic Map