Assay Details
| Gene Symbol: TWN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101328.4 | NP_172913.1 | 16 - 17 | 2340 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TWN2| Gene Symbol: TWN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101328.4 | NP_172913.1 | 16 - 17 | 2340 | 67 | |
Set Membership: |
|