Assay Details
Gene Symbol: DL1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101352.3 | NP_172936.1 | 1 - 2 | 409 | 109 | |
GenBank mRNA | AJ304841.1 | - | 1 - 2 | 333 | 109 | |
AY039955.1 | - | 1 - 2 | 294 | 109 | ||
AY150494.1 | - | 1 - 2 | 133 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
