Assay Details
Gene Symbol: DL1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101352.3 | NP_172936.1 | 5 - 6 | 734 | 91 | |
GenBank mRNA | AJ304841.1 | - | 5 - 6 | 658 | 91 | |
AY039955.1 | - | 5 - 6 | 619 | 91 | ||
AY150494.1 | - | 5 - 6 | 458 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
