Assay Details
Gene Symbol: DL1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101352.3 | NP_172936.1 | 7 - 8 | 881 | 150 | |
GenBank mRNA | AJ304841.1 | - | 7 - 8 | 805 | 150 | |
AY039955.1 | - | 7 - 8 | 766 | 150 | ||
AY150494.1 | - | 7 - 8 | 605 | 150 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
