Assay Details
| Gene Symbol: NUDT18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101355.3 | NP_172939.1 | 1 - 2 | 397 | 65 | |
| GenBank mRNA | AK117453.1 | - | 1 - 2 | 325 | 65 | |
| BT005432.1 | - | 1 - 2 | 74 | 65 | ||
More Information
Set Membership: |
|

NUDT18
Genomic Map