Assay Details
| Gene Symbol: NUDT18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101355.3 | NP_172939.1 | 3 - 4 | 582 | 110 | |
| GenBank mRNA | AK117453.1 | - | 3 - 4 | 510 | 110 | |
| BT005432.1 | - | 3 - 4 | 259 | 110 | ||
More Information
Set Membership: |
|

NUDT18
Genomic Map