Assay Details
| Gene Symbol: SWI3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102019.4 | NP_173589.1 | 5 - 6 | 1708 | 61 | |
| GenBank mRNA | AY091026.1 | - | 5 - 6 | 1679 | 61 | |
| AY117245.1 | - | 5 - 6 | 1520 | 61 | ||
More Information
Set Membership: |
|

SWI3C
Genomic Map