Assay Details
| Gene Symbol: SWI3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102019.4 | NP_173589.1 | 7 - 8 | 1864 | 67 | |
| GenBank mRNA | AY091026.1 | - | 7 - 8 | 1835 | 67 | |
| AY117245.1 | - | 7 - 8 | 1676 | 67 | ||
More Information
Set Membership: |
|

SWI3C
Genomic Map