Assay Details
| Gene Symbol: SWI3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102019.4 | NP_173589.1 | 8 - 9 | 2034 | 77 | |
| GenBank mRNA | AY091026.1 | - | 8 - 9 | 2005 | 77 | |
| AY117245.1 | - | 8 - 9 | 1846 | 77 | ||
More Information
Set Membership: |
|

SWI3C
Genomic Map