Assay Details
Gene Symbol: TRZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106128.3 | NP_177608.2 | 2 - 3 | 675 | 111 | |
GenBank mRNA | AJ428989.1 | - | 2 - 3 | 344 | 111 | |
BT025035.1 | - | 2 - 3 | 344 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
