Assay Details
Gene Symbol: TRZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106128.3 | NP_177608.2 | 5 - 6 | 959 | 65 | |
GenBank mRNA | AJ428989.1 | - | 5 - 6 | 628 | 65 | |
BT025035.1 | - | 5 - 6 | 628 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
