Assay Details
Gene Symbol: TRZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106128.3 | NP_177608.2 | 6 - 7 | 1021 | 96 | |
GenBank mRNA | AJ428989.1 | - | 6 - 7 | 690 | 96 | |
BT025035.1 | - | 6 - 7 | 690 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
