Assay Details
Gene Symbol: SmD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106280.4 | NP_177757.1 | 4 - 5 | 410 | 75 | |
GenBank mRNA | AF380651.1 | - | 4 - 5 | 339 | 75 | |
AY054149.1 | - | 4 - 5 | 324 | 75 | ||
AY087251.1 | - | 4 - 5 | 397 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
