Assay Details
| Gene Symbol: SKP2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198486.2 | NP_001185415.1 | 2 - 3 | 505 | 79 | |
| NM_001334757.1 | NP_001322101.1 | 2 - 3 | 380 | 79 | ||
| NM_001334758.1 | NP_001322102.1 | 1 - 2 | 613 | 79 | ||
| NM_106351.4 | NP_565147.1 | 1 - 2 | 613 | 79 | ||
| GenBank mRNA | AY087441.1 | - | 1 - 2 | 267 | 79 | |
| BT024747.1 | - | 1 - 2 | 211 | 79 | ||
More Information
Set Membership: |
|

SKP2B
Genomic Map