Assay Details
| Gene Symbol: AT1G77090 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106359.5 | NP_565149.2 | 2 - 3 | 476 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT1G77090| Gene Symbol: AT1G77090 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106359.5 | NP_565149.2 | 2 - 3 | 476 | 66 | |
Set Membership: |
|