Assay Details
| Gene Symbol: ADH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106362.3 | NP_177837.1 | 6 - 7 | 1275 | 102 | |
| GenBank mRNA | AY045612.1 | - | 6 - 7 | 1076 | 102 | |
| AY090330.1 | - | 6 - 7 | 1021 | 102 | ||
More Information
Set Membership: |
|

ADH1
Genomic Map