Assay Details
| Gene Symbol: AT1G77310 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334775.1 | NP_001321460.1 | 5 - 6 | 1092 | 74 | |
| NM_106380.7 | NP_177855.4 | 6 - 7 | 1176 | 74 | ||
More Information
Set Membership: |
|

AT1G77310
Genomic Map