Assay Details
| Gene Symbol: AT1G77330 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106382.3 | NP_565154.1 | 1 - 2 | 299 | 92 | |
| GenBank mRNA | AK228946.1 | - | 1 - 2 | 199 | 92 | |
| BT026444.1 | - | 1 - 2 | 109 | 92 | ||
More Information
Set Membership: |
|

AT1G77330
Genomic Map