Assay Details
Gene Symbol: BGAL16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334779.1 | NP_001320598.1 | 11 - 12 | 1637 | 84 | |
NM_001334780.1 | NP_001320597.1 | 12 - 13 | 1559 | 84 | ||
NM_001334781.1 | NP_001320596.1 | 9 - 10 | 1133 | 84 | ||
NM_106391.2 | NP_177866.2 | 12 - 13 | 1544 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
