Assay Details
Gene Symbol: BGAL16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334779.1 | NP_001320598.1 | 2 - 3 | 378 | 76 | |
NM_001334780.1 | NP_001320597.1 | 2 - 3 | 378 | 76 | ||
NM_106391.2 | NP_177866.2 | 2 - 3 | 378 | 76 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001334779.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
