Assay Details
Gene Symbol: BGAL16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334779.1 | NP_001320598.1 | 6 - 7 | 976 | 81 | |
NM_001334780.1 | NP_001320597.1 | 7 - 8 | 898 | 81 | ||
NM_001334781.1 | NP_001320596.1 | 4 - 5 | 472 | 81 | ||
NM_106391.2 | NP_177866.2 | 7 - 8 | 883 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
