Assay Details
| Gene Symbol: TAPX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334784.1 | NP_001321064.1 | 1 - 2 | 330 | 68 | |
| NM_106398.3 | NP_177873.1 | 1 - 2 | 372 | 68 | ||
| GenBank mRNA | AK229693.1 | - | 1 - 2 | 248 | 68 | |
| AY085554.1 | - | 1 - 2 | 262 | 68 | ||
More Information
Set Membership: |
|

TAPX
Genomic Map