Assay Details
| Gene Symbol: PDIL1-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106400.3 | NP_177875.1 | 9 - 10 | 1386 | 88 | |
| GenBank mRNA | AK226862.1 | - | 9 - 10 | 1329 | 88 | |
More Information
Set Membership: |
|

PDIL1-2
Genomic Map