Assay Details
| Gene Symbol: AT1G77680 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106417.3 | NP_177891.1 | 5 - 6 | 3057 | 132 | |
| GenBank mRNA | AK229034.1 | - | 5 - 6 | 3026 | 132 | |
More Information
Set Membership: |
|

AT1G77680
Genomic Map