Assay Details
| Gene Symbol: LAX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106418.4 | NP_177892.1 | 2 - 3 | 457 | 131 | |
| GenBank mRNA | AY054266.1 | - | 2 - 3 | 379 | 131 | |
| BT008300.1 | - | 2 - 3 | 356 | 131 | ||
More Information
Set Membership: |
|

LAX3
Genomic Map