Assay Details
Gene Symbol: GGH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106514.4 | NP_177988.1 | 5 - 6 | 832 | 95 | |
GenBank mRNA | AF389281.1 | - | 5 - 6 | 722 | 95 | |
BT000554.1 | - | 5 - 6 | 683 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
