Assay Details
| Gene Symbol: APC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106524.2 | NP_565188.1 | 2 - 3 | 885 | 101 | |
| GenBank mRNA | BT033023.1 | - | 2 - 3 | 383 | 101 | |
More Information
Set Membership: |
|

APC6
Genomic Map