Assay Details
Gene Symbol: VHA-A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036222.3 | NP_001031299.1 | 11 - 12 | 1125 | 67 | |
NM_106539.6 | NP_178011.1 | 12 - 13 | 1084 | 67 | ||
GenBank mRNA | AK230248.1 | - | 12 - 13 | 1353 | 67 | |
AK317358.1 | - | 11 - 12 | 1082 | 67 | ||
AY059909.1 | - | 12 - 13 | 985 | 67 | ||
AY081296.1 | - | 12 - 13 | 985 | 67 | ||
AY085759.1 | - | 12 - 13 | 985 | 67 | ||
BT002589.1 | - | 11 - 12 | 860 | 67 | ||
BT008383.1 | - | 11 - 12 | 860 | 67 | ||
U65638.1 | - | 11 - 12 | 888 | 67 |
Gene Symbol: AT5G21020 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK230328.1 | - | 1376 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
