Assay Details
| Gene Symbol: VHA-A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036222.3 | NP_001031299.1 | 8 - 9 | 894 | 90 | |
| NM_106539.6 | NP_178011.1 | 9 - 10 | 853 | 90 | ||
| GenBank mRNA | AK230248.1 | - | 9 - 10 | 1122 | 90 | |
| AK317358.1 | - | 8 - 9 | 851 | 90 | ||
| AY059909.1 | - | 9 - 10 | 754 | 90 | ||
| AY081296.1 | - | 9 - 10 | 754 | 90 | ||
| AY085759.1 | - | 9 - 10 | 754 | 90 | ||
| BT002589.1 | - | 8 - 9 | 629 | 90 | ||
| BT008383.1 | - | 8 - 9 | 629 | 90 | ||
| U65638.1 | - | 8 - 9 | 657 | 90 | ||
| Gene Symbol: AT5G21020 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK230328.1 | - | 1607 | 90 | ||
More Information
Set Membership: |
|

VHA-A
Genomic Map