Assay Details
| Gene Symbol: HAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198504.2 | NP_001185433.1 | 5 - 6 | 1467 | 107 | |
| NM_106550.4 | NP_565197.3 | 5 - 6 | 1583 | 107 | ||
More Information
Set Membership: |
|

HAC1
Genomic Map