Assay Details
Gene Symbol: SYP52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036224.2 | NP_001031301.1 | 5 - 6 | 703 | 89 | |
NM_001334895.1 | NP_001321707.1 | 4 - 5 | 728 | 89 | ||
NM_106607.5 | NP_565213.1 | 4 - 5 | 808 | 89 | ||
GenBank mRNA | AF355756.1 | - | 4 - 5 | 697 | 89 | |
BT003941.1 | - | 4 - 5 | 679 | 89 | ||
BT006106.1 | - | 3 - 4 | 521 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
